Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
para
Entrez Gene ID
32619
Gene Name
paralytic
Gene Aliases
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285944
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042815.2 NP_001036280.1 16 - 17 2558 70
NM_001042816.2 NP_001036281.1 16 - 17 2558 70
NM_001201671.1 NP_001188600.1 16 - 17 2558 70
NM_001201672.1 NP_001188601.1 16 - 17 2558 70
NM_001201673.1 NP_001188602.1 15 - 16 2495 70
NM_001201674.1 NP_001188603.1 15 - 16 2525 70
NM_001201678.1 NP_001188607.1 15 - 16 2495 70
NM_001201680.1 NP_001188609.1 15 - 16 2495 70
NM_001201682.1 NP_001188611.1 16 - 17 2558 70
NM_001201685.1 NP_001188614.1 15 - 16 2495 70
NM_001201687.1 NP_001188616.1 16 - 17 2558 70
NM_001201688.1 NP_001188617.1 15 - 16 2495 70
NM_001201693.1 NP_001188622.1 16 - 17 2486 70
NM_001201695.2 NP_001188624.1 16 - 17 2486 70
NM_001201696.1 NP_001188625.1 14 - 15 2462 70
NM_001201697.1 NP_001188626.1 16 - 17 2558 70
NM_001201698.1 NP_001188627.1 15 - 16 2525 70
NM_001201699.1 NP_001188628.1 14 - 15 2462 70
NM_001201700.1 NP_001188629.1 15 - 16 2525 70
NM_001201701.1 NP_001188630.1 14 - 15 2462 70
NM_001201702.1 NP_001188631.1 15 - 16 2525 70
NM_001201703.3 NP_001188632.1 16 - 17 3048 70
NM_001201704.1 NP_001188633.1 14 - 15 2462 70
NM_001201705.1 NP_001188634.1 15 - 16 2453 70
NM_001201706.1 NP_001188635.1 16 - 17 2558 70
NM_001201708.1 NP_001188637.1 15 - 16 2495 70
NM_001201710.1 NP_001188639.1 12 - 13 2241 70
NM_001201711.1 NP_001188640.1 14 - 15 2396 70
NM_001201712.1 NP_001188641.1 13 - 14 2333 70
NM_001201713.1 NP_001188642.1 13 - 14 2333 70
NM_001201715.1 NP_001188644.1 14 - 15 2390 70
NM_001201717.1 NP_001188646.1 15 - 16 2525 70
NM_001201718.1 NP_001188647.1 14 - 15 2462 70
NM_001201719.1 NP_001188648.1 15 - 16 2525 70
NM_001201720.1 NP_001188649.1 15 - 16 2525 70
NM_001201722.1 NP_001188651.1 15 - 16 2525 70
NM_001272690.1 NP_001259619.1 16 - 17 2558 70
NM_001298403.1 NP_001285332.1 16 - 17 2711 70
NM_001298404.1 NP_001285333.1 15 - 16 2522 70
NM_078647.4 NP_523371.2 16 - 17 2558 70

View Full Product Details