Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or22c
- Entrez Gene ID
-
33381
- Gene Name
- Odorant receptor 22c
- Gene Aliases
-
22C.1, 22c, CG15377, DOR16, DOR22C.1, Dmel\CG15377, Dmel_CG15377, Or22C.1
- Chromosome Location
-
Chr.2L: 2054589 - 2057967 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026396
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078730.3 |
NP_523454.2 |
1 - 2 |
528 |
74 |
View Full Product Details