Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp
Entrez Gene ID
33941
Gene Name
Thrombospondin
Gene Aliases
CG11326, CT31613, D-TSP, DTSP, Dmel\CG11326, Dmel_CG11326, SP295, TSP, tsp
Chromosome Location
Chr.2L: 6686791 - 6709086 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031850
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144330.2 NP_001137802.1 13 - 14 3679 89
NM_001144331.2 NP_001137803.1 4 - 5 2099 89
NM_001273219.1 NP_001260148.1 3 - 4 2151 89
NM_078771.3 NP_523495.2 12 - 13 4014 89

View Full Product Details