The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001299564.1 | NP_001286493.1 | 4 - 5 | 811 | 66 |
NM_001299565.1 | NP_001286494.1 | 5 - 6 | 964 | 66 | |
NM_057848.3 | NP_477196.1 | 5 - 6 | 1390 | 66 | |
NM_166184.2 | NP_725594.1 | 5 - 6 | 1165 | 66 | |
NM_166185.2 | NP_725595.1 | 5 - 6 | 866 | 66 | |
GenBank mRNA | AY060397.1 | - | 5 - 6 | 1372 | 66 |