Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- hppy
- Entrez Gene ID
-
37203
- Gene Name
- happyhour
- Gene Aliases
-
CG7097, CG7097/hppy, Dmel\CG7097, Dmel_CG7097, EP-704, GLK/KHS1, Hppy, MAP4K3, anon-WO0153538.68, l(2)SH1261, l(2)SH2 1261
- Chromosome Location
-
Chr.2R: 19178738 - 19227193 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0263395
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166335.3 |
NP_725863.1 |
8 - 9 |
1503 |
85 |
View Full Product Details