Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nahoda
Entrez Gene ID
37641
Gene Name
CG12781-PA
Gene Aliases
CG12781, CT36793, Dmel\CG12781, Dmel_CG12781
Chromosome Location
Chr.2R: 22896174 - 22916719 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034797
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259545.2 NP_001246474.1 6 - 7 3075 54
NM_079091.4 NP_523815.2 6 - 7 3075 54
NM_166569.3 NP_726264.1 6 - 7 2607 54
GenBank mRNA AY060632.1 - 6 - 7 3032 54

View Full Product Details