Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp4e3
- Entrez Gene ID
-
34291
- Gene Name
- Cytochrome P450-4e3
- Gene Aliases
-
4e3, CG4105, CYP4E3, Cyp4E3, Dmel\CG4105, Dmel_CG4105, P-450, P450, cyp4e3
- Chromosome Location
-
Chr.2L: 9747839 - 9750071 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015035
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078803.2 |
NP_523527.1 |
1 - 2 |
214 |
73 |
GenBank mRNA |
BT030822.1 |
- |
1 - 2 |
214 |
73 |
View Full Product Details