Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RpL7
Entrez Gene ID
34352
Gene Name
Ribosomal protein L7
Gene Aliases
BEST:GH04411, CG4897, Dmel\CG4897, Dmel_CG4897, L7, RPL7, Rp L7, rpL7, rpl7
Chromosome Location
Chr.2L: 10201004 - 10202321 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005593
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298865.1 NP_001285794.1 2 - 3 439 62
NM_078807.3 NP_523531.1 2 - 3 343 62
GenBank mRNA AY089648.1 - 2 - 3 341 62

View Full Product Details