Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hgo
Entrez Gene ID
34552
Gene Name
homogentisate 1,2-dioxygenase
Gene Aliases
CG4779, Dmel\CG4779, Dmel_CG4779
Chromosome Location
Chr.2L: 11136393 - 11138565 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040211
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078820.3 NP_523544.2 7 - 8 1224 82
GenBank mRNA BT003496.1 - 7 - 8 1222 82

View Full Product Details