Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rho-6
- Entrez Gene ID
-
34640
- Gene Name
- rhomboid-6
- Gene Aliases
-
CG17212, Dmel\CG17212, Dmel_CG17212, Rho-related [33C1], rho 6, rho6
- Chromosome Location
-
Chr.2L: 12130516 - 12133411 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032415
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176024.3 |
NP_788038.1 |
2 - 3 |
459 |
67 |
View Full Product Details