Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rho-6
Entrez Gene ID
34640
Gene Name
rhomboid-6
Gene Aliases
CG17212, Dmel\CG17212, Dmel_CG17212, Rho-related [33C1], rho 6, rho6
Chromosome Location
Chr.2L: 12130516 - 12133411 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032415
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298943.1 NP_001285872.1 3 - 4 633 63
NM_176024.3 NP_788038.1 3 - 4 643 63

View Full Product Details