Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tep4
Entrez Gene ID
35248
Gene Name
Thioester-containing protein 4
Gene Aliases
CG10363, CT29102, Dmel\CG10363, Dmel_CG10363, TEP IV, TEP4, TEP4mel, TEPIV, Tep IV, TepIV, dTEPIV, tep4, tepIV
Chromosome Location
Chr.2L: 19548507 - 19556451 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041180
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273660.1 NP_001260589.1 6 - 7 2308 73
NM_078879.3 NP_523603.2 5 - 6 2285 73

View Full Product Details