Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lar
Entrez Gene ID
35259
Gene Name
Leukocyte-antigen-related-like
Gene Aliases
CG10443, DLAR, DLar, Dlar, Dmel\CG10443, Dmel_CG10443, LAR, PTP38A, Ptp38A, dlar, lar
Chromosome Location
Chr.2L: 19586623 - 19732069 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000464
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259165.1 NP_001246094.1 8 - 9 1397 61
NM_078880.3 NP_523604.2 7 - 8 1351 61
NM_165313.4 NP_724220.2 8 - 9 1693 61

View Full Product Details