Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsp42Eg
Entrez Gene ID
35616
Gene Name
Tetraspanin 42Eg
Gene Aliases
BcDNA:GM06962, CG12142, Dm.Tsp42Eg, Dmel\CG12142, Dmel_CG12142
Chromosome Location
Chr.2R: 7028911 - 7033233 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033128
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078909.4 NP_523633.1 1 - 2 248 66
GenBank mRNA AF274017.1 - 1 - 2 230 66
AY113403.1 - 1 - 2 246 66

View Full Product Details