Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CSN7
- Entrez Gene ID
-
35816
- Gene Name
- COP9 signalosome subunit 7
- Gene Aliases
-
CG2038, CH7, CSN7b, Csn7, Dch7, Dmel\CG2038, Dmel_CG2038, csn7
- Chromosome Location
-
Chr.2R: 8421033 - 8428577 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028836
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136535.3 |
NP_610379.2 |
2 - 3 |
520 |
88 |
GenBank mRNA |
BT150080.1 |
- |
2 - 3 |
507 |
88 |
View Full Product Details