Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rme-8
Entrez Gene ID
35939
Gene Name
Receptor mediated endocytosis 8
Gene Aliases
CG8014, Dmel\CG8014, Dmel_CG8014, E(shi)2-1, l(2)45Ba, rme-8
Chromosome Location
Chr.2R: 9167763 - 9177261 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015477
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299307.1 NP_001286236.1 12 - 13 2870 80
NM_136623.2 NP_610467.1 12 - 13 2873 80

View Full Product Details