Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cpr47Ef
- Entrez Gene ID
-
36194
- Gene Name
- Cuticular protein 47Ef
- Gene Aliases
-
CG13214, DmelCpr47Ef, Dmel\CG13214, Dmel_CG13214
- Chromosome Location
-
Chr.2R: 11266915 - 11272575 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033603
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136816.3 |
NP_610660.2 |
13 - 14 |
1666 |
110 |
View Full Product Details