Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl
Entrez Gene ID
37757
Gene Name
egalitarian
Gene Aliases
CG4051, Dmel\CG4051, Dmel_CG4051, EGL, Egl, egal
Chromosome Location
Chr.2R: 23734904 - 23746829 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000562
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299850.1 NP_001286779.1 3 - 4 615 75
NM_166623.3 NP_726360.3 3 - 4 1158 75

View Full Product Details