Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pur-alpha
Entrez Gene ID
43797
Gene Name
Purine-rich binding protein-alpha
Gene Aliases
CG1507, Dmel\CG1507, Dmel_CG1507, PURalpha, Pur alpha, Pur-a, dPur alpha, pur-alpha, purA
Chromosome Location
Chr.4: 558016 - 567257 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022361
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272132.1 NP_001259061.1 4 - 5 613 146
NM_001272133.1 NP_001259062.1 4 - 5 430 146
NM_001272134.1 NP_001259063.1 4 - 5 438 146
NM_079885.2 NP_524624.1 4 - 5 536 146
NM_166767.2 NP_726579.1 4 - 5 533 146
NM_166768.2 NP_726580.1 4 - 5 458 146
NM_166769.2 NP_726581.1 4 - 5 438 146
GenBank mRNA AY061185.1 - 4 - 5 505 146
BT126365.1 - 4 - 5 438 146

View Full Product Details