Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PlexA
Entrez Gene ID
43832
Gene Name
Plexin A
Gene Aliases
BcDNA:GM05237, CG11081, D-Plex A, DPlexA, Dmel\CG11081, Dmel_CG11081, Plex1, PlexA1, PlexinA, lincRNA.927, plex, plex A, plexA
Chromosome Location
Chr.4: 1009895 - 1027101 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025741
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258499.2 NP_001245428.1 7 - 8 6016 131
NM_079898.3 NP_524637.2 7 - 8 5874 131
NM_166804.4 NP_726627.1 6 - 7 4435 131
NM_166805.4 NP_726628.1 7 - 8 4711 131

View Full Product Details