Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Edem1
Entrez Gene ID
31154
Gene Name
ER degradation enhancer, mannosidase alpha-like 1
Gene Aliases
CG3810, DM-pI, Dmel\CG3810, Dmel_CG3810, EDEM1, EG.86E4.2, EG:86E4.2, dEDEM1, edem1
Chromosome Location
Chr.X: 1903704 - 1908827 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023511
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258554.2 NP_001245483.1 3 - 4 3362 89
NM_166915.2 NP_726778.1 4 - 5 3678 89
GenBank mRNA AY119600.1 - 4 - 5 3678 89

View Full Product Details