Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Unc-76
Entrez Gene ID
31179
Gene Name
Uncoordinated 76
Gene Aliases
BcDNA:GH10260, CG18851, CG3981, Dmel\CG3981, Dmel_CG3981, Dunc-76, EG:67A9.1, UNC-76, l(1)G0066, l(1)G0158, l(1)G0310, l(1)G0333, l(1)G0360, unc-76, unc76
Chromosome Location
Chr.X: 2080610 - 2093948 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040395
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272243.1 NP_001259172.1 4 - 5 1842 69
NM_001272244.1 NP_001259173.1 4 - 5 1063 69
NM_130625.3 NP_569981.1 4 - 5 1842 69
NM_166927.2 NP_726792.1 5 - 6 1732 69
GenBank mRNA AY069376.1 - 4 - 5 1812 69

View Full Product Details