Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or56a
- Entrez Gene ID
-
37269
- Gene Name
- Odorant receptor 56a
- Gene Aliases
-
56E.1, 56a, CG12501, DOR59, DmOr56a, Dmel\CG12501, Dmel_CG12501, OR56a, Or-56a
- Chromosome Location
-
Chr.2R: 19769461 - 19771233 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034473
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079072.2 |
NP_523796.2 |
3 - 4 |
949 |
128 |
View Full Product Details