Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qkr58E-2
Entrez Gene ID
37562
Gene Name
quaking related 58E-2
Gene Aliases
CG5821, Dmel\CG5821, Dmel_CG5821, qkr58e-2, qrk58E-2
Chromosome Location
Chr.2R: 22415560 - 22419001 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022985
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299795.1 NP_001286724.1 1 - 2 811 61
NM_001299796.1 NP_001286725.1 1 - 2 562 61
NM_079086.3 NP_523810.2 1 - 2 562 61
GenBank mRNA BT021292.1 - 1 - 2 558 61

View Full Product Details