Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or59a
- Entrez Gene ID
-
37711
- Gene Name
- Odorant receptor 59a
- Gene Aliases
-
59D.1, 59a, AN6, CG9820, DOR46, DOR59D.1, Dmel\CG9820, Dmel_CG9820, OR59a, Or46, Or59D.1, dor46
- Chromosome Location
-
Chr.2R: 23439868 - 23441065 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026384
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079097.1 |
NP_523821.1 |
1 - 2 |
727 |
70 |
View Full Product Details