Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NaCP60E
Entrez Gene ID
37981
Gene Name
Na channel protein 60E
Gene Aliases
CG18506, CG34405, CG9071, CT24831, DIC60, DSC, DSC1, DSCI, Dmel\CG34405, Dmel_CG34405, Na-CP, dsc, dsc1, smi60E
Chromosome Location
Chr.2R: 24891614 - 24921677 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202078.3 NP_001189007.2 10 - 11 3056 83
NM_001259578.1 NP_001246507.1 7 - 8 2157 83
NM_001259579.3 NP_001246508.1 10 - 11 3319 83
NM_001259580.2 NP_001246509.1 10 - 11 3056 83
NM_001259581.2 NP_001246510.1 10 - 11 3319 83
NM_001274243.2 NP_001261172.1 10 - 11 3056 83
NM_001274244.2 NP_001261173.1 10 - 11 3056 83
NM_001274245.2 NP_001261174.1 9 - 10 3113 83
NM_001274246.2 NP_001261175.1 10 - 11 3056 83
NM_166696.5 NP_726495.3 10 - 11 3319 83

View Full Product Details