Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or67b
- Entrez Gene ID
-
39120
- Gene Name
- Odorant receptor 67b
- Gene Aliases
-
67B.2, 67b, CG14176, DmOr67b, Dmel\CG14176, Dmel_CG14176
- Chromosome Location
-
Chr.3L: 9627316 - 9629193 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036019
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079283.5 |
NP_524007.2 |
9 - 10 |
1209 |
67 |
View Full Product Details