Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp69a
- Entrez Gene ID
-
39411
- Gene Name
- Odorant-binding protein 69a
- Gene Aliases
-
69a, CG10436, DmelObp69a, Dmel\CG10436, Dmel_CG10436, PB-PRP1, PBP1, PBPRP-1, Pbprp1, pbprp-1, pbprp1
- Chromosome Location
-
Chr.3L: 12396171 - 12396957 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011279
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079315.2 |
NP_524039.2 |
3 - 4 |
275 |
130 |
View Full Product Details