Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RpS4
Entrez Gene ID
39484
Gene Name
Ribosomal protein S4
Gene Aliases
CG11276, Dmel\CG11276, Dmel_CG11276, RPS4, Rp S4, SX4, anon-EST:Posey270, rpS4
Chromosome Location
Chr.3L: 13041756 - 13044234 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011284
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300126.1 NP_001287055.1 4 - 5 465 71
NM_079329.3 NP_524053.2 3 - 4 564 71
NM_168537.2 NP_729871.1 4 - 5 465 71
GenBank mRNA BT011369.1 - 4 - 5 465 71

View Full Product Details