Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pex13
Entrez Gene ID
36462
Gene Name
Peroxin 13
Gene Aliases
CG4663, DmelPex13, Dmel\CG4663, Dmel_CG4663, PEX13, pex13
Chromosome Location
Chr.2R: 13228276 - 13230234 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033812
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137006.3 NP_610850.1 2 - 3 1184 62
GenBank mRNA BT001726.1 - 2 - 3 1178 62

View Full Product Details