Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DJ-1alpha
- Entrez Gene ID
-
36543
- Gene Name
- CG6646-PA
- Gene Aliases
-
CG6646, DJ-1, DJ-1A, DJ-1a, Dj-1alpha, Dmel\CG6646, Dmel_CG6646, dDJ-1alpha
- Chromosome Location
-
Chr.2R: 13942534 - 13943377 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033885
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137072.1 |
NP_610916.1 |
1 - 2 |
200 |
67 |
View Full Product Details