Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Arc1
- Entrez Gene ID
-
36595
- Gene Name
- Activity-regulated cytoskeleton associated protein 1
- Gene Aliases
-
Arc, CG12505, Dmel\CG12505, Dmel_CG12505, anon-WO0153538.31, anon-WO0153538.40, anon-WO0153538.41, anon-WO0153538.43, anon-WO0153538.45, arc1, dARC1, dArc1, darc1
- Chromosome Location
-
Chr.2R: 14357927 - 14360297 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033926
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137111.3 |
NP_610955.1 |
1 - 1 |
2296 |
102 |
GenBank mRNA |
AY061464.1 |
- |
1 - 1 |
2267 |
102 |
View Full Product Details