Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sin1
Entrez Gene ID
36604
Gene Name
SAPK-interacting protein 1
Gene Aliases
CG10105, Dmel\CG10105, Dmel_CG10105, dSIN1, dSin1, sin1
Chromosome Location
Chr.2R: 14411825 - 14413851 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033935
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137119.4 NP_610963.1 1 - 1 1952 101
GenBank mRNA BT003261.1 - 1 - 1 1913 101

View Full Product Details