The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_137142.5 | NP_610986.1 | 2 - 3 | 557 | 70 |
GenBank mRNA | AY060730.1 | - | 2 - 3 | 540 | 70 |