Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hr51
- Entrez Gene ID
-
36702
- Gene Name
- Hormone receptor 51
- Gene Aliases
-
CG16801, CG16801/NR2E3, DHR51, Dmel\CG16801, Dmel_CG16801, HR51, NR2E3, Nr2e3, PNR, UNF, fdx, unf
- Chromosome Location
-
Chr.2R: 15331325 - 15339384 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034012
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137188.3 |
NP_611032.2 |
5 - 6 |
989 |
68 |
GenBank mRNA |
BT122090.2 |
- |
5 - 6 |
713 |
68 |
View Full Product Details