Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp4aa1
- Entrez Gene ID
-
36752
- Gene Name
- CG8302-PB
- Gene Aliases
-
4aa1, CG8302, CYP4AA1, Cyp4AA1, Dmel\CG8302, Dmel_CG8302
- Chromosome Location
-
Chr.2R: 15942864 - 15947266 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034053
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137223.3 |
NP_611067.2 |
2 - 3 |
411 |
80 |
View Full Product Details