Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
norpA
Entrez Gene ID
31376
Gene Name
no receptor potential A
Gene Aliases
CG3620, CdkA, DIP2, Dmel\CG3620, Dmel_CG3620, MRE18, NORPA, NorpA, PI-PLC, PLC, PLC-beta, PLCbeta, anon-EST:Posey221, nofC, rof, x12, x13, x16, x24
Chromosome Location
Chr.X: 4322626 - 4365408 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262738
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169190.2 NP_001162661.1 7 - 8 1719 60
NM_080330.4 NP_525069.2 5 - 6 988 60
NM_167008.2 NP_726925.1 6 - 7 1450 60
GenBank mRNA AF181641.1 - 5 - 6 971 60

View Full Product Details