Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
axo
Entrez Gene ID
43923
Gene Name
axotactin
Gene Aliases
CG13715, CG13717, CG18296, CG32234, CG42664, CG43225, CT1277, Dmel\CG43225, Dmel_CG43225
Chromosome Location
Chr.3L: 4629604 - 4687317 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262870
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259701.1 NP_001246630.1 15 - 16 4836 61
NM_001259702.1 NP_001246631.1 15 - 16 4836 61
NM_001259703.1 NP_001246632.1 15 - 16 4836 61
NM_001274493.1 NP_001261422.1 15 - 16 4945 61
NM_001274494.1 NP_001261423.1 15 - 16 4945 61
NM_001274495.1 NP_001261424.1 15 - 16 4945 61
NM_079917.5 NP_524656.5 15 - 16 4945 61

View Full Product Details