Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CysRS
- Entrez Gene ID
-
36784
- Gene Name
- Cysteinyl-tRNA synthetase
- Gene Aliases
-
Aats-cys, BcDNA:LD21177, CG8431, CRS, Dmel\CG8431, Dmel_CG8431, dCARS
- Chromosome Location
-
Chr.2R: 16143147 - 16145906 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027091
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137243.3 |
NP_611087.1 |
4 - 5 |
1590 |
61 |
GenBank mRNA |
AF132160.1 |
- |
4 - 5 |
1562 |
61 |
View Full Product Details