Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nopo
Entrez Gene ID
37083
Gene Name
no poles
Gene Aliases
CG5140, Dmel\CG5140, Dmel_CG5140, NOPO, Nopo, TRIP
Chromosome Location
Chr.2R: 18180496 - 18182519 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034314
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274142.1 NP_001261071.1 4 - 5 963 66
NM_137461.4 NP_611305.1 4 - 5 842 66
GenBank mRNA AY060610.1 - 4 - 5 809 66

View Full Product Details