Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sun
- Entrez Gene ID
-
44046
- Gene Name
- stunted
- Gene Aliases
-
ATP5E, ATPE, ATPsynepsilon, BcDNA:RE19513, BcDNA:RH48911, CG9032, Dmel\CG9032, Dmel_CG9032, Sun, l(1)13Fe, l(1)XV
- Chromosome Location
-
Chr.X: 15847859 - 15849258 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014391
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167468.3 |
NP_727905.1 |
3 - 4 |
306 |
83 |
GenBank mRNA |
BT099753.1 |
- |
3 - 4 |
288 |
83 |
View Full Product Details