Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fim
Entrez Gene ID
32721
Gene Name
Fimbrin
Gene Aliases
CG8649, Dmel\CG8649, Dmel_CG8649, FIM, fim
Chromosome Location
Chr.X: 17285704 - 17301929 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024238
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167565.2 NP_728074.1 1 - 2 415 84
GenBank mRNA BT133379.1 - 1 - 2 389 84

View Full Product Details