Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hydra
Entrez Gene ID
33058
Gene Name
CG1338-PA
Gene Aliases
CG1338, Dmel\CG1338, Dmel_CG1338
Chromosome Location
Chr.X: 20670832 - 20677063 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031128
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169341.2 NP_001162812.2 2 - 3 365 69
NM_001258852.1 NP_001245781.1 2 - 3 364 69
NM_001258853.1 NP_001245782.1 2 - 3 382 69
NM_001258854.1 NP_001245783.1 2 - 3 380 69
NM_001258855.1 NP_001245784.1 2 - 3 381 69
NM_001258856.1 NP_001245785.1 2 - 3 371 69
NM_134561.7 NP_608405.4 2 - 3 504 69
NM_167711.3 NP_728358.1 2 - 3 346 69

View Full Product Details