Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fwd
Entrez Gene ID
45374
Gene Name
four wheel drive
Gene Aliases
CG7004, Dmel\CG7004, Dmel_CG7004, Fwd, PI4K, PI4K3B, PI4KIIIbeta, anon-WO0118547.259
Chromosome Location
Chr.3L: 305393 - 319884 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004373
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274274.1 NP_001261203.1 5 - 6 2335 62
NM_001370043.1 NP_001356971.1 4 - 5 2568 62
NM_080083.4 NP_524822.1 4 - 5 2568 62
NM_167826.2 NP_728518.1 4 - 5 1812 62
NM_167827.2 NP_728519.1 3 - 4 641 62
GenBank mRNA AF242375.2 - 3 - 4 641 62
AY052049.1 - 4 - 5 2549 62

View Full Product Details