Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
O-fut2
Entrez Gene ID
31098
Gene Name
O-fucosyltransferase 2
Gene Aliases
CG14789, Dmel\CG14789, Dmel_CG14789, EG:BACN32G11.6, O-FUT2, O-fut, OFut2, Ofut2
Chromosome Location
Chr.X: 1464363 - 1465996 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027791
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130560.4 NP_569916.1 1 - 1 1581 80
GenBank mRNA AY047568.1 - 1 - 1 1591 80

View Full Product Details