Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk9
Entrez Gene ID
37498
Gene Name
pickpocket 9
Gene Aliases
CG13489, CG13490, CG34369, Dmel\CG34369, Dmel_CG34369
Chromosome Location
Chr.2R: 21964173 - 21970664 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085398
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299771.1 NP_001286700.1 2 - 3 730 86
NM_137778.2 NP_611622.2 2 - 3 730 86

View Full Product Details