Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcp1
Entrez Gene ID
37925
Gene Name
TFIIF-interacting CTD phosphatase
Gene Aliases
21483550, CG12252, DmFcp1, Dmel\CG12252, Dmel_CG12252, Dmfcp1, fcp1
Chromosome Location
Chr.2R: 24567987 - 24571821 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035026
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299917.1 NP_001286846.1 2 - 3 803 81
NM_138090.2 NP_611934.1 2 - 3 642 81

View Full Product Details