Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APC7
- Entrez Gene ID
-
31594
- Gene Name
- Anaphase Promoting Complex subunit 7
- Gene Aliases
-
Apc7, CG14444, Dmel\CG14444, Dmel_CG14444
- Chromosome Location
-
Chr.X: 6597088 - 6599699 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029879
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132101.4 |
NP_572329.3 |
1 - 2 |
110 |
65 |
View Full Product Details