Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP2
Entrez Gene ID
252479
Gene Name
DISCO Interacting Protein 2
Gene Aliases
CG7020, Ddip2, Dmel\CG7020, Dmel_CG7020
Chromosome Location
Chr.3L: 250229 - 257954 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024806
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259601.2 NP_001246530.1 12 - 13 4231 67
NM_001274262.1 NP_001261191.1 12 - 13 4234 67
NM_001274263.1 NP_001261192.1 12 - 13 4282 67
NM_001274264.1 NP_001261193.1 12 - 13 4267 67
NM_138175.4 NP_612019.2 12 - 13 4285 67

View Full Product Details