Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mwh
Entrez Gene ID
38131
Gene Name
multiple wing hairs
Gene Aliases
BP1018, CG13913, CG34022, CG43772, Dmel\CG43772, Dmel_CG43772
Chromosome Location
Chr.3L: 1200786 - 1232700 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264272
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274314.1 NP_001261243.1 6 - 7 1737 68
NM_001274315.1 NP_001261244.1 6 - 7 1740 68
NM_138238.3 NP_612082.1 4 - 5 1228 68
GenBank mRNA AM231270.1 - 4 - 5 1213 68
BT023932.1 - 4 - 5 1225 68

View Full Product Details